missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BLOTTO and Tween 20 in TBS

Blocks non-specific binding site for antibodies on the membrane

Brand:  Alfa Aesar™ J62341.AP

Code : NEW

Additional Details : Weight : 0.00050kg

 View more versions of this product

Product Code. 15494739

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Blocks non-specific binding site for antibodies on the membrane
Blocking Buffer
−30° to −10°C
Liquid
500mL
Videos
SDS