Alfa Aesar™ BLOTTO and Tween 20 in TBS, with 0.02% sodium azide

Blocking reagent for immunohistochemistry experiments

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J63026.AP

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00050kg



Product Code. 15425299

Quantity Price
1 £ 31.06 / 500mL
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Specification

Storage Requirements −30° to −10°C
Product Type Blocking Buffer
Format Liquid
Applications Blocking reagent for immunohistochemistry experiments
Quantity 500mL
Documentation

Technical documents