missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BLOTTO and Tween 20 in PBS, with 0.02% sodium azide

Blocking reagent for immunohistochemistry experiments

Brand:  Alfa Aesar™ J62304.AP

Code : NEW

Additional Details : Weight : 0.00050kg

 View more versions of this product

Product Code. 15474709

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Blocking reagent for immunohistochemistry experiments
Blocking Buffer
−30° to −10°C
Liquid
500mL
Videos
SDS