missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BLOTTO in PBS

Blocking reagent for immunohistochemistry experiments

Brand:  Alfa Aesar™ J60296.AK

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15453089

  • £31.06 / 250mL


Estimated Shipment date
from Supplier 08-06-2017
Add to basket

Specifications Description & Specifications

Specifications

Blocking reagent for immunohistochemistry experiments
Blocking Buffer
−30° to −10°C
Liquid
250mL
Documentation

Technical Documents