Alfa Aesar™ BLOTTO Antifoam in PBS

Blocking buffer for immunohistochemistry experiments

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J63851.AK

250ML BLOTTO antifoam in PBS

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg



Product Code. 15456009

Quantity Price
1 £ 31.06 / 250mL
Estimated Shipment date
from Supplier 02-02-2017
Add to basket


Description and Specification

Specification

Format Liquid
Product Type Blocking Buffer
Applications Blocking buffer for immunohistochemistry experiments
Quantity 250mL
Storage Requirements −30° to −10°C