Alfa Aesar™ BLOTTO antifoam in PBS, with 0.02% sodium azide

Blocking reagent for immunohistochemistry experiments

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J61888.AK

250ML BLOTTO antifoam in PBS, with 0.02% sodium azide

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg



Product Code. 15404379

Quantity Price
1 £ 31.06 / 250mL
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Specification

Quantity 250mL
Format Liquid
Product Type Blocking Buffer
Applications Blocking reagent for immunohistochemistry experiments
Storage Requirements −30° to −10°C