missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ BLOTTO antifoam in PBS, with 0.02% sodium azide
Blocking reagent for immunohistochemistry experiments
Brand: Alfa Aesar™ J61888.AP
Code : NEW
Additional Details : Weight : 0.00050kg
Specifications
Blocking reagent for immunohistochemistry experiments | |
Blocking Buffer | |
−30° to −10°C |
Liquid | |
500mL |