missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BLOTTO

Blocking reagent for immunohistochemistry experiments

Brand:  Alfa Aesar™ J61417.AP

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00050kg

 View more versions of this product

Product Code. 15444029

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Blocking reagent for immunohistochemistry experiments.
Blocking Buffer
−30° to −10°C
Liquid
500mL
Documentation

Technical Documents