Learn More
Blood Group Antigen H (O) Type 1, clone: 17-206, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9810-80
Code : Z2
Additional Details : Weight : 0.01000kg
Description
The H antigen is a precursor to the ABO blood group antigens, present in people of all common blood types. The Bombay phenotype (hh) does not express antigen H on red blood cells, and therefore this type will also lack A or B antigens, similar to the O blood group. However, unlike O group, the H antigen is absent, hence the individuals produce isoantibodies to antigen H as well as to both A and B antigens. If they receive blood from someone with O blood group, the anti-H antibodies will bind to the H antigen on the red blood cells of the donor blood and destroy the RBCs by complement-mediated lysis. Therefore, people with Bombay phenotype can receive blood only from other hh donors, although they can donate as though they were type O. Some individuals with the blood group A1 may also be able to produce anti-H antibodies due to the complete conversion of all the H antigen to A1 antigen. Production of the H antigen, or its deficiency in the Bombay phenotype, is controlled at the H locus on chromosome 19. The H locus contains three exons that span more than 5 kb of genomic DNA, and encodes the fucosyltransferase that produces the H antigen on RBCs. The H antigen is a carbohydrate sequence with carbohydrates linked mainly to protein (with a minor fraction attached to ceramide moiety).
Description: The monoclonal antibody 17-206 recognizes the human blood group antigen H (O) type 1, a subtype primarily found in epithelial and secretory cells. The precursor H antigen is modified by addition of saccharides to carbohydrate chains of proteins and lipids on erythrocytes and epithelial cells. The type, attachment, location, and sequence of saccharides determines the blood group antigen subtype and is mediated by multiple glycosyl transferases. Depending on ABO blood type, the H antigen is converted into either the A antigen, B antigen, both the A and B antigen, or left unconverted (O antigen). Evidence that the blood group antigen H is not expressed in tumor cells and that the level correlates with disease progression suggests that the blood group antigen H may be involved in cell proliferation, cell adhesion, and angiogenesis, although its exact function remains to be determined. The 17-206 antibody does not recognize blood group antigen H (O) type 2.Applications Reported: This 17-206 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections and ELISA.Applications Tested: This 17-206 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using either high or low pH antigen retrieval at less than or equal to 5 ug/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.Specifications
Blood Group Antigen H (O) Type 1 | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-8°C. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
17-206 | |
Unconjugated | |
ABH, BG-H1, BG-H, Blood Group H ab antigen | |
Affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgG3 |
For Research Use Only.