Learn More
BD Vacutainer Blood collection research kit P700 contains 20 x blood collection tubes for basic metabolic research, 10 x tubes/foil pouch & 2 x foil pouches/kit Becton Dickinson
Brand:  BD Vacutainer 366473
Blood collection research kit P700 contains 20 x blood collection tubes for basic metabolic research, 10 x tubes/foil pouch & 2 x foil pouches/kit Becton Dickinson
Code : Z1
Additional Details : Weight : 0.50000kg
Product Code. 12804781