missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters-Mixed Blocks

Catalog Number 15377948
Click to view available options
No. of Places:
1
3
18
30
32
This item is not returnable. View return policy
This item is not returnable. View return policy

Constructed of aluminum alloy

A variety of blocks for any application

TRUSTED_SUSTAINABILITY
Capacity (Metric) 30 x 0.5 mL, 20 x 0.2 mL
For Use With (Equipment) Thermo Scientific Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Material Aluminum Alloy
No. of Places 30
Type Block
Includes 0.5 mL and 0.2 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'