missing translation for 'onlineSavingsMsg'
Learn More

Blimp-1 Rat anti-Mouse, PE, Clone: 5E7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9850-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15597236

  • £242.00 / 100µg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression.
Specifications

Specifications

Blimp-1
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q60636
BLIMP 1, B Lymphocyte Induced Maturation Protein 1, BLIMP1, Positive Regulatory Domain I Binding Factor 1, PR Domain Containing 1, PRDI BF1, PRDM1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
5E7
PE
Prdm1
Liquid
Prdm1
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12142
Videos
Documents

For Research Use Only.