missing translation for 'onlineSavingsMsg'
Learn More

Blimp-1, clone: 6D3, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5963-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227637

  • £154.31 / 100Āµg


EU Stock 3
Add to basket

Description

Description

Description: This 6D3 monoclonal antibody reacts with B lymphocyte-induced maturation protein-1 (Blimp-1), a transcriptional repressor expressed in B and T cells. This approximately 90-kDa protein contains five consecutive Kruppel-like zinc finger domains by which it binds DNA. In B cells, Blimp-1 promotes terminal differentiation into antibody-secreting plasma cells during a primary immune response. In addition, Blimp-1 has been implicated in germinal center formation in response to T-dependent and T-independent antigens, as well as in some diffuse large B cell lymphomas. Expressed in CD4+ and CD8+ effector/memory T cells and FoxP3+ regulatory T cells, reports have suggested possible roles for Blimp-1 in T cell homeostasis and suppression of IL-2 production. Moreover, by suppressing Th1 differentiation, Blimp-1 plays a role in T helper cell differentiation. Finally, Blimp-1 has been demonstrated to play a significant role in the terminal differentiation of effector and effector memory CD8+ T cells during viral infection.Applications Reported: This 6D3 antibody has been reported for use in immunoblotting (WB) and immunohistology staining of paraffin-embedded tissue sections.Applications Tested: This 6D3 antibody has been tested by western blot of reduced A20 cell lysates. For western blottiing, this antibody can be used at 1-10 μg/mL. Moreover, this antibody was tested by immunohistochemistry of formalin-fixed paraffin embedded human tonsil tissue. Antigen retrieval was performed using 10mM citrate buffer at pH 6.0. For immunohistochemistry, this antibody can be used at 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Blimp1
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q60636, O75626
BLIMP 1, B Lymphocyte Induced Maturation Protein 1, BLIMP1, Positive Regulatory Domain I Binding Factor 1, PR Domain Containing 1, PRDI BF1, PRDM1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
ImmunohistochemistryĆ¾Western Blot
6D3
Unconjugated
PRDM1
Purified
Prdm1
IgG2a
100μg
Store at 2-8°C.
Primary
639
Videos

For Research Use Only.