missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Blaubrand™ ETERNA™ Class A Volumetric Flasks

Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks are calibrated to contain a precise amount of liquid at a specific temperature. Narrow neck and wide neck options available in a variety of sizes. Includes polypropylene stopper and DE-M marking.

Brand:  Brand™ 36847

Code : 03

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11884000

  • £45.40 / Pack of 2


Estimated Shipment
19-06-2018
Add to basket

Description

Description

  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • Includes polypropylene stopper and DE-M marking
Specifications

Specifications

Calibrated To Contain
Class A
12/21
Clear
Borosilicate Glass
±0.06mL
2Pack
USP individual and USP batch-certificate, DIN EN ISO 1042
Polypropylene
Stopper
50mL
Yes
Narrow
Videos