missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Blaubrand™ Trapezoidal Volumetric Flask with Polypropylene Stopper

Manufactured from Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Trapezoidal Volumetric Flask with Polypropylene Stopper prevents tipping and provides stability with flat surface.

Brand:  Brand™ 36408

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11878341

  • £35.17 / Pack of 2


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Manufactured from Borosilicate Glass.
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Includes batch number and batch certificate.
  • DAkkS calibration certificate available upon request.
  • Flat surface prevents tipping and provides stability.
  • Includes polypropylene stopper.
Specifications

Specifications

Trapezoidal
DIN EN ISO 1042, USP and DAkkS calibration certificates available upon request.
Polypropylene
Clear
Borosilicate Glass
±0.06mL
±0.06mL
Calibrated To Contain
Class A
12/21
50mL
Yes
2Piece
Videos