missing translation for 'onlineSavingsMsg'
Learn More

CellPath BiTran™ High Visibility Barrier Film Specimen Bag

Designed for transporting wet tissue from one place to another, and for the short to long-term storage of tissue.

Brand:  CellPath WAA-0300-12A

Code : LB

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12310649

  • £443.00 / Pack of 250
Estimated Shipment: 03-05-2024
to see stock.
Add to basket

Description

Description

  • Re-sealable specimen bags
  • Manufactured from a special barrier film with a unique double zip lock closure
  • Barrier film is impermeable to formalin vapour, preventing hazardous vapour polluting the laboratory once the bag is sealed
  • Air is expelled from the bag by squeezing prior to closure
  • Double zip lock seal provides a leak proof closure that can be easily opened and closed as many times as required allowing the specimen to be re-examined whenever necessary
Specifications

Specifications

Specimen Bag
For transporting wet tissue, short to long-term storage of tissue
250 Pack
Zip Seal
High-visibility Barrier Film
150 x 150mm (6 x 6 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers