missing translation for 'onlineSavingsMsg'
Learn More

(±)-Bisoprolol hemifumarate, Tocris Bioscience™

β1 antagonist

Brand:  Tocris (Bio-Techne) 091450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13428153

  • £317.52 / 50mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A selective β1-adrenergic antagonist. Has a Kd of 2-3 nM at the β1 receptor and a β1/β2 selectivity of approximately 100-fold.
Specifications

Chemical Identifiers

104344-23-2
441.521
bisoprolol fumarate, 1-4-2-isopropoxyethoxy methyl phenoxy-3-isopropylamino propan-2-ol fumarate, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy-2-propanol, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy propan-2-ol
but-2-enedioic acid;1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
C22H35NO8
RZPZLFIUFMNCLY-UHFFFAOYSA-N
53394791
CC(C)NCC(COC1=CC=C(C=C1)COCCOC(C)C)O.C(=CC(=O)O)C(=O)O

Specifications

50mg
104344-23-2
bisoprolol fumarate, 1-4-2-isopropoxyethoxy methyl phenoxy-3-isopropylamino propan-2-ol fumarate, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy-2-propanol, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy propan-2-ol
CC(C)NCC(COC1=CC=C(C=C1)COCCOC(C)C)O.C(=CC(=O)O)C(=O)O
441.521
383.48
(±)-Bisoprolol hemifumarate
C22H35NO8
RZPZLFIUFMNCLY-UHFFFAOYSA-N
but-2-enedioic acid;1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
53394791
>98%
Videos
Documents