missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth titanium isopropoxide, 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Color | Yellow |
Packaging | ChemSeal™ bottles |
Linear Formula | BiTi[OCH(CH3)2]7 |
Quantity | 25 mL |
UN Number | UN1219 |
Sensitivity | Moisture Sensitive |
Formula Weight | 670.48 |
Physical Form | Liquid |
Chemical Name or Material | Bismuth titanium isopropoxide |
Safety and Handling
Product Identifier
- Bismuth titanium isopropoxide, 5% w/v in isopropanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Bismuth titanium isopropoxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'