missing translation for 'onlineSavingsMsg'
Learn More

Bismuth shot, 1-5mm (0.04-0.20in), 99.999% (metal basis), Thermo Scientific Chemicals

Catalog Number 11318947
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.999% (metals basis)
Quantity 10 g
Chemical Name or Material Bismuth shot

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'