missing translation for 'onlineSavingsMsg'
Learn More

Bismuth powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11381158
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]
This item is not returnable. View return policy
7440-69-9
Bi
208.98
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
5359367
CHEBI:33301
bismuth
[Bi]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3089
Quantity 500 g
Percent Purity 99.5%
Mesh Size −325 Mesh
Odor Odorless
Weight Apparent density: 3.2g/cm3 to 4.0g/cm3
Form Powder
Assay (metals basis)
Chemical Name or Material Bismuth powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'