Learn More
Bismuth Internal Standard for method ASTM D5059 Part A and C, Bi @ 0.793g/L, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045338.22
72.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth Internal Standard for method ASTM D5059 | |
100 g | |
MFCD00134033 |
Part A and C, Bi at 0.793g/L | |
Brown | |
Liquid |
RUO – Research Use Only