Learn More
Bismuth Indium Tin ingot (Field's metal), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046895.A3
Code : NEW
Additional Details : Weight : 2.00000kg
Description
Bismuth Indium Tin ingot (Field's metal) is used for die casting and rapid prototyping. It acts as an alternative to Wood's metal which is an eutectic alloy of 50% bismuth, 26.7% lead, 13.3% tin, and 10% cadmium by weight.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth Indium Tin ingot (Field's metal) | |
2 kg | |
Ingot |
Bi:In:Sn; 32.5:51:16.5 wt% | |
Insoluble in water. |
RUO – Research Use Only