missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
In semiconductor research, multilayer thin-film, magnetoresistant film.Bismuth(III) selenide is used as a semiconductor and thermoelectric material. It is also used in semiconductor research. Further, it finds applications magnetoresistant film and semiconductor research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 710°C |
Color | Gray |
Physical Form | Pieces |
Quantity | 10 g |
UN Number | UN3283 |
Merck Index | 14,1278 |
Solubility Information | Insoluble in water; Decomposes when heated in air; Decomposes in nitric acid and aqua regiaSoluble in strong acids; Insoluble in water and organic solvents. |
Formula Weight | 654.84 |
Percent Purity | 99.999% |
Density | 6.82 g/mL |
Show More |
Safety and Handling
Product Identifier
- Bismuth(III) selenide
- Danger
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P403-Store in a well-ventilated place.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'