missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) oxide, typically 99.99% (metals basis), Thermo Scientific Chemicals
34.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Used as an adhesive and sealant chemical, flame retardant, pigment, propellant and blowing agent. It is also used as a solvent. Other uses are in batteries, electrical and electronic products, explosive materials, ink, toner, and colorant products, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ca 1890°C |
Melting Point | 817°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Insoluble in water. Soluble in acids. |
Formula Weight | 465.96 |
Density | 8.9 |
Chemical Name or Material | Bismuth(III) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'