missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048479
Click to view available options
Quantity:
50 g
250 g
1304-76-3
Bi2O3
465.96
MFCD00003462
TYIXMATWDRGMPF-UHFFFAOYSA-N
dibismuth(3+) trioxidandiide
[O--].[O--].[O--].[Bi+3].[Bi+3]
1304-76-3
Bi2O3
465.96
MFCD00003462
TYIXMATWDRGMPF-UHFFFAOYSA-N
dibismuth(3+) trioxidandiide
[O--].[O--].[O--].[Bi+3].[Bi+3]

It is used as a precursor to the preparation of other compounds of bismuth, chemical reagents. It has specialized uses in optical glass, flame-retardant paper, in glaze formulations as a substitute for lead oxides. It is used as solid electrolyte because of its high thermal expansion property. It is used in radio frequency shielding, compound for catheters and other medical tubing products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼1890°C
Melting Point 817°C
Color Yellow
Physical Form Powder
Quantity 50 g
Merck Index 14,1273
Solubility Information Soluble in acids. Insoluble in water & organic solvents.
Formula Weight 465.96
Percent Purity 99.9995%
Grade Puratronic™
Density 8.9 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'