Learn More
Bismuth(III) oxide, nanopowder, 99.9%, Thermo Scientific Chemicals
Description
Bismuth(III) oxide is used in the preparation of BiFeO3perovskite nanoparticles. It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generators. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ∼1890°C |
Melting Point | 817°C |
Color | Yellow |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,1273 |
Solubility Information | Soluble in acids. Insoluble in water. |
Formula Weight | 465.96 |
Percent Purity | 99.9% |
Density | 8.9 g/mL |
Show More |
RUO – Research Use Only