missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog Number p-4509913
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
1304-76-3
Bi2O3
465.96
MFCD00003462
TYIXMATWDRGMPF-UHFFFAOYSA-N
Bismuth trioxide
dibismuth(3+) trioxidandiide
[O--].[O--].[O--].[Bi+3].[Bi+3]
1304-76-3
Bi2O3
465.96
MFCD00003462
TYIXMATWDRGMPF-UHFFFAOYSA-N
Bismuth trioxide
dibismuth(3+) trioxidandiide
[O--].[O--].[O--].[Bi+3].[Bi+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1890°C
Melting Point 825.0°C
Color Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9%
Linear Formula Bi2O3
Fieser 01,59
Merck Index 15,1273
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 465.95
Percent Purity 99.9%
Grade Extra Pure
Loss on Drying 0.2% max.
Packaging Plastic bottle
Specific Gravity 8.9
Density 8.9000g/mL
Chemical Name or Material Bismuth(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'