missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) nitrate pentahydrate, ACS, 98% min, Thermo Scientific Chemicals

Catalog Number 11321589
Click to view available options
Quantity:
250 g
1 kg
10035-06-0
BiH10N3O14
485.07
MFCD00149157
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
160911
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
10035-06-0
BiH10N3O14
485.07
MFCD00149157
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate
160911
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Luminous paints, preparation of bismuth saltsBismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30°C (decomposition)
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 98.0%
Solubility Information Soluble in dilute HNO3 acid solution. Decomposes in water to subnitrate. Soluble in dilute acetic acid,glycerol,acetone. Insoluble in alcohol,ethyl acetateSoluble in acetone,acetic acid and glycerol. Insoluble in ethanol and ethyl acetate.
IUPAC Name bismuth(3+) pentahydrate trinitrate
Formula Weight 485.07 (394.99 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Appearance Crystalline
Density 2.83 g/cm3
Chemical Name or Material Bismuth (III) Nitrate Pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P220-Keep/Store away from clothing/combustible materials.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'