missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) nitrate pentahydrate, ACS, 98% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Luminous paints, preparation of bismuth saltsBismuth(III) nitrate pentahydrate is an oxidant for a variety of 4-substituted Hantzsch 1,4-dihydropyridines. It also serves as a reagent for selective oxidation of sufides to sulfoxides. Further, it is involved in the preparation of Dragendorff reagent and used as a TLC stain. It is also used to prepare other bismuth compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 30°C (decomposition) |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | 98.0% |
Solubility Information | Soluble in dilute HNO3 acid solution. Decomposes in water to subnitrate. Soluble in dilute acetic acid,glycerol,acetone. Insoluble in alcohol,ethyl acetateSoluble in acetone,acetic acid and glycerol. Insoluble in ethanol and ethyl acetate. |
IUPAC Name | bismuth;trinitrate;pentahydrate |
Formula Weight | 485.07 (394.99 Anhydrous) |
Grade | ACS |
Sensitivity | Hygroscopic |
Appearance | Crystalline |
Show More |
Safety and Handling
Signal Word
- Danger
- Oxidising solid Category 2
- Serious eye damage/eye irritation Category 2
- H372-Causes damage to organs.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'