missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) nitrate pentahydrate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 80.0°C |
Melting Point | 30.0°C |
Quantity | 500 g |
Linear Formula | Bi(NO3)3·5H2O |
Merck Index | 15,1271 |
Solubility Information | Solubility in water: may decompose |
IUPAC Name | bismuth;trinitrate;pentahydrate |
Formula Weight | 485.07 |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Show More |
Safety and Handling
Product Identifier
- Bismuth(III) nitrate pentahydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'