missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048477
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
7787-64-6
BiI3
589.69
MFCD00010894
KOECRLKKXSXCPB-UHFFFAOYSA-K
bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
111042
[I-].[I-].[I-].[Bi+3]
This item is not returnable. View return policy
7787-64-6
BiI3
589.69
MFCD00010894
KOECRLKKXSXCPB-UHFFFAOYSA-K
bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
111042
[I-].[I-].[I-].[Bi+3]

Polycrystalline films of bismuth triiodide are used as beta direct and digital detectors for medical imaging applications. Bismuth(III) iodide is used as an ice nucleant. It is also used in electron microscopy to reveal synapses. Addition of bismuth triiodide has been shown to improve the glass forming ability and thermal stability of Ge-Te-BiI(3) chalcogenide glasses. Solution-processable BiI3 nanosheets have been shown to exhibit the potential to be used as HTLs (Hole Transport Layer) in conventional OPV (organic photovoltaic) devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼500°C
Melting Point 408°C
Color Gray to Black
Physical Form Lump/Powder
Quantity 2 g
UN Number UN3260
Merck Index 14,1269
Solubility Information Insoluble in water. Highly soluble in ethanol,benzene,toluene and xylene,hydrochloric acid and in hydriodic acid.
IUPAC Name triiodobismuthane
Formula Weight 589.69
Percent Purity 99.999%
Grade Puratronic™
Odor Acidic
Sensitivity Hygroscopic; Light sensitive
Density 5.778 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth(III) iodide
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) iodide, Puratronic
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'