missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bismuth(III) fluoride, 99.999% (metals basis)

Brand:  Alfa Aesar™ 011844.22

Code : NEW

Additional Details : CAS Number : 7787-61-3 Weight : 0.10000kg Transport : UN number : 3260 Chem class : 8 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11381908

  • £662.61 / 100g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

Bismuth(III) fluoride
265.98
UN3260
7787-61-3
MFCD00010893
14,1265

Specifications

Bismuth(III) fluoride
Powder
(metals basis)
MFCD00010893
14,1265
Odorless
Hygroscopic
100g
7787-61-3
99.999%
265.98
UN3260
−200 Mesh
1.74
727°C
Videos
SDS