missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride, 97+% (dry wt.), may cont. up to 3% water, Thermo Scientific Chemicals

Catalog Number 11458763
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
7787-60-2
BiCl3
315.33
MFCD00003461
JHXKRIRFYBPWGE-UHFFFAOYSA-K
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
24591
trichlorobismuthane
Cl[Bi](Cl)Cl
This item is not returnable. View return policy
7787-60-2
BiCl3
315.33
MFCD00003461
JHXKRIRFYBPWGE-UHFFFAOYSA-K
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
24591
trichlorobismuthane
Cl[Bi](Cl)Cl

Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

Solubility
Soluble in hydrochloric acid, nitric acid, diethyl ether, ethyl acetate and acetone. Insoluble in alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point ∼430°C (sublimation)
Melting Point 230°C to 232°C
Quantity 25 g
UN Number UN3260
Merck Index 14,1262
Solubility Information Soluble in hydrochloric acid,nitric acid,diethyl ether,ethyl acetate and acetone. Insoluble in alcohol.
Formula Weight 315.34
Percent Purity 98%
Concentration or Composition (by Analyte or Components) May contain up to 3% water
Odor Irritating
Sensitivity Moisture sensitive
Density 4.75
Assay (dry wt.)
Chemical Name or Material Bismuth (III) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'