missing translation for 'onlineSavingsMsg'
Learn More

Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min, Thermo Scientific Chemicals

Catalog Number p-7051109
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1304-85-4
MFCD00064844
Bismuth subnitrate
This item is not returnable. View return policy
1304-85-4
MFCD00064844
Bismuth subnitrate

Ceramic glazes and enamel fluxes

It is used pharmaceutically in wound dressing, ointment component and as an antiacid. In cosmetics it is used as a cosmetic white pigment. Used in Ceramic glazes and enamel fluxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
UN Number UN1477
Merck Index 14,1284
Solubility Information Practically insoluble in water and alcohol; Soluble in dilute HNO3 and HCl
Formula Weight 1461.98
Concentration or Composition (by Analyte or Components) Bi2O3 79% min.
Odor Odorless
Sensitivity Hygroscopic
Density 6.04 g/mL
Chemical Name or Material Bismuth hydroxide nitrate oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bismuth hydroxide nitrate oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'