missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bismuth hexafluoro-2,4-pentanedionate, 97+%

Brand:  Alfa Aesar™ 041201.03

Code : NEW

Additional Details : CAS Number : 142617-56-9 Weight : 0.00100kg


Quantity 1g

 View more versions of this product

Product Code. 11389557

  • £49.70 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket


Specifications

Chemical Identifiers

Bismuth hexafluoro-2,4-pentanedionate
142617-56-9
830.14
MFCD00270924

Specifications

Bismuth hexafluoro-2,4-pentanedionate
Powder
830.14
110°C (0.05mmHg) (subl.)
1g
142617-56-9
≥97%
MFCD00270924
≈188°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bismuth hexafluoro-2,4-pentanedionate

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS