missing translation for 'onlineSavingsMsg'
Learn More

Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049144
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
12233-56-6
MFCD00064629
This item is not returnable. View return policy
12233-56-6
MFCD00064629

Bismuth germanium oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 1245.68
Percent Purity 99.9995%
Grade Puratronic™
Odor Odorless
Assay (metals basis)
Chemical Name or Material Bismuth germanium oxide
compliance-icons
Product Identifier
  • Bismuth germanium
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'