missing translation for 'onlineSavingsMsg'
Learn More

Bismuth aluminum oxide hydrate, 95.5%, Thermo Scientific™

Catalog Number p-7051070
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
MFCD00798526
Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate
This item is not returnable. View return policy
MFCD00798526
Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water; Soluble in nitric acid; Typical tap density 0;69g/cm3
Formula Weight 771.84 (Anhydrous)
Percent Purity 95.5%
Chemical Name or Material Bismuth aluminum oxide hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'