Alfa Aesar™ Bismuth 2-ethylhexanoate monoisopropoxide, 99.9% (metals basis), 5% w/v in isopropanol


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 039148.AE

BIS(ETHYLHEXYLOXY)BISMUTH ISOPROPOXIDE 5% W IPA 0,

UNSPSC: 12352103

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg


Quantity 100mL

Product Code. 11363518

Quantity Price
1 £ 259.17 / 100mL
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Bismuth 2-ethylhexanoate monoisopropoxide
Formula Weight 554.49
MDL Number MFCD00210603
UN Number UN1219
Synonym Bismuth 2-ethylhexano-isopropoxide

Specification

Formula Weight 554.49
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bismuth 2-ethylhexanoate monoisopropoxide
Quantity 100mL
Concentration 5% w/v in isopropanol
Physical Form Liquid
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00210603
UN Number UN1219
Synonym Bismuth 2-ethylhexano-isopropoxide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H361-H319-H336
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Color Colorless to pale yellow
Flash Point 12° (54°F)
Odor Alcohol-like
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS H Statement
H225-H361-H319-H336
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.