missing translation for 'onlineSavingsMsg'
Learn More

Bismuth 2-ethylhexanoate, 99.99+%, 70% w/v in xylene, Thermo Scientific™

Catalog Number p-7051553
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
67874-71-9
C24H48BiO6
641.622
MFCD00043803
MWXWNYHYPMRCHN-UHFFFAOYSA-N
2-Ethylhexanoic acid bismuth salt
131857451
bismuth;2-ethylhexanoic acid
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]
This item is not returnable. View return policy
67874-71-9
C24H48BiO6
641.622
MFCD00043803
MWXWNYHYPMRCHN-UHFFFAOYSA-N
2-Ethylhexanoic acid bismuth salt
131857451
bismuth;2-ethylhexanoic acid
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN1307
Formula Weight 638.61
Percent Purity ≥99.99%
Physical Form Liquid
Chemical Name or Material Bismuth 2-ethylhexanoate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bismuth 2-ethylhexanoate 70% w/v in xylene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P262-Do not get in eyes, on skin, or on clothing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'