missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bismuth 2-ethylhexanoate, 99.99+%, 70% w/v in xylene

Brand:  Alfa Aesar™ 041521.14

Code : NEW

Additional Details : CAS Number : 67874-71-9 Weight : 0.02500kg Transport : UN number : 1307 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11324268

  • £118.10 / 25g


Estimated Shipment date
from Supplier 23-04-2018
Add to basket

Specifications

Chemical Identifiers

Bismuth 2-ethylhexanoate
638.61
UN1307
67874-71-9
MFCD00043803
2-Ethylhexanoic acid bismuth salt

Specifications

Bismuth 2-ethylhexanoate
70% w/v in xylene
Liquid
638.61
UN1307
25g
67874-71-9
≥99.99%
MFCD00043803
2-Ethylhexanoic acid bismuth salt
Videos
SDS