missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth 2-ethylhexanoate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Bismuth 2-ethylhexanoate is used as an industrial lubricant and dessicant. It is also used for synthesizing of urethane foam, urethane coating and other urethane products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 5 g |
Solubility Information | Not miscible with water. |
Formula Weight | 638.61 |
Physical Form | Viscous Liquid |
Chemical Name or Material | Bismuth 2-ethylhexanoate |
Safety and Handling
Product Identifier
- Bismuth 2-ethylhexanoate
- Warning
- Reproductive toxicity Category 2
- H361d-Suspected of damaging the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'