missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisindolylmaleimide 1, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
25 mg
Description
Bisindolylmaleimide is used as a highly selective inhibitor of all PKC isoforms and slight inhibitor of GSK-3, used to selectively probe for PKC-mediated pathways for transduction of hormone, cytokine, and growth factor signals, very potent and selective inhibitor of protein kinase C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bisindolylmaleimide is used as a highly selective inhibitor of all PKC isoforms and slight inhibitor of GSK-3, used to selectively probe for PKC-mediated pathways for transduction of hormone, cytokine, and growth factor signals, very potent and selective inhibitor of protein kinase C.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Bisindolylmaleimide is used as a highly selective inhibitor of all PKC isoforms and slight inhibitor of GSK-3, used to selectively probe for PKC-mediated pathways for transduction of hormone, cytokine, and growth factor signals, very potent and selective inhibitor of protein kinase C.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Color | Orange |
Quantity | 5 mg |
Solubility Information | Insoluble in water. |
Formula Weight | 412.48 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Bisindolylmaleimide 1 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'