missing translation for 'onlineSavingsMsg'
Learn More

Bisabolene, mixture of isomers, Thermo Scientific Chemicals

Catalog Number p-7024306
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
495-62-5
C15H24
204.357
MFCD00129080
XBGUIVFBMBVUEG-CCEZHUSRSA-N
e-gamma-bisabolene, unii-l8m7r52o3b, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, e, 1e-bisabola-1 10 ,4,7 11-triene 4e-4-1,5-dimethylhex-4-en-1-ylidene-1-methylcyclohexene, gamma-bisabolene, e, gamma-bisabolene, 4e, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, 4e, .gamma.-bisabolene, e gamma-bisabolene, 1-methyl-4-6-methylhept-5-en-2-ylidene cyclohex-1-ene
3033866
CHEBI:49238
(4Z)-1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohexene
CC1=CCC(=C(C)CCC=C(C)C)CC1
This item is not returnable. View return policy
495-62-5
C15H24
204.357
MFCD00129080
XBGUIVFBMBVUEG-CCEZHUSRSA-N
e-gamma-bisabolene, unii-l8m7r52o3b, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, e, 1e-bisabola-1 10 ,4,7 11-triene 4e-4-1,5-dimethylhex-4-en-1-ylidene-1-methylcyclohexene, gamma-bisabolene, e, gamma-bisabolene, 4e, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, 4e, .gamma.-bisabolene, e gamma-bisabolene, 1-methyl-4-6-methylhept-5-en-2-ylidene cyclohex-1-ene
3033866
CHEBI:49238
(4Z)-1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohexene
CC1=CCC(=C(C)CCC=C(C)C)CC1

It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

Solubility
Soluble in alcohol, water, (0.008994 mg/L @ 25°C (est)).

Notes
Air Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 0.89
Refractive Index 1.494
Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol,water,(0.008994mg/L @ 25°C (est)).
Formula Weight 204.36
Chemical Name or Material Bisabolene, mixture of isomers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'