missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BIS-TRIS propane, 0.2M buffer soln., pH 6.5

Catalog Number p-7042388
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
64431-96-5
C11H26N2O6
282.337
MFCD00004689
HHKZCCWKTZRCCL-UHFFFAOYSA-N
125132
CHEBI:40947
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
This item is not returnable. View return policy
64431-96-5
C11H26N2O6
282.337
MFCD00004689
HHKZCCWKTZRCCL-UHFFFAOYSA-N
125132
CHEBI:40947
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration 0.2 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material BIS-TRIS propane buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'