missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)palladium(II) chloride, 98%, ACROS Organics™

Brand:  Acros Organics 299250250

Code : 29

Additional Details : CAS Number : 13965-03-2 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10649263

  • £378.10 / 25g


Estimated Shipment
04-07-2018
Add to basket


Specifications

Chemical Identifiers

Bis(triphenylphosphine)palladium(II) chloride
98%
[(C6H5)3P]2PdCl2
MFCD00009593
98%
C36H30Cl2P2Pd
701.89
Dichlorobis(triphenylphosphine)palladium(II)

Specifications

Bis(triphenylphosphine)palladium(II) chloride
Authentic
Liquid
98%
[(C6H5)3P]2PdCl2
MFCD00009593
06,59; 07,95; 08,151; 17,111
Dichlorobis(triphenylphosphine)palladium(II)
13965-03-2
Glass bottle
98%
C36H30Cl2P2Pd
701.89
25g
Clear colorless
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bis(triphenylphosphine)palladium(II) chloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS