missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(triphenylphosphine)palladium(II) chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 260°C |
Color | Colorless |
Infrared Spectrum | Authentic |
Assay Percent Range | 14.8 to 15.5% (Pd) |
Packaging | Glass bottle |
Linear Formula | [(C6H5)3P]2PdCl2 |
Quantity | 100 g |
Fieser | 06,59; 07,95; 08,151; 17,111 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 701.89 |
Show More |
Safety and Handling
Product Identifier
- Bis(triphenylphosphine)palladium(II) chloride
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H413-May cause long lasting harmful effects to aquatic life.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'