missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)palladium(II) diacetate, 99%, Thermo Scientific Chemicals

Catalog Number 10511081
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14588-08-0
C40H36O4P2Pd
749.09
MFCD00010013
YOUIUHDWQIUKAO-UHFFFAOYSA-L
bis triphenylphosphinepalladium acetate
73357379
palladium(2+);triphenylphosphanium;diacetate
CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
14588-08-0
C40H36O4P2Pd
749.09
MFCD00010013
YOUIUHDWQIUKAO-UHFFFAOYSA-L
bis triphenylphosphinepalladium acetate
73357379
palladium(2+);triphenylphosphanium;diacetate
CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 136.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 14% min. (Pd)
Packaging Glass bottle
Linear Formula [(C6H5)3P]2Pd(O2CCH3)2
Quantity 5 g
Formula Weight 749.08
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bis(triphenylphosphine)palladium(II) diacetate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'