missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)nickel(II)chloride, 98%, ACROS Organics™

Brand:  Acros Organics 217500500

Code : 29

Additional Details : CAS Number : 14264-16-5 Weight : 0.10000kg Transport : UN number : 2923 Chem class : 1a Pack group : III

Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10723221

  • £80.20 / 50g


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis(triphenylphosphine)nickel(II)chloride
14264-16-5
C36H30Cl2NiP2
654.2
Dichlorobis(triphenylphosphine)nickel(II)
98%
98%
[(C6H5)3P]2NiCl2
MFCD00009592

Specifications

Bis(triphenylphosphine)nickel(II)chloride
14264-16-5
Authentic
Crystals or Powder
98%
> 300.0°C
654.2
50g
Dark green to dark gray
98%
97.5% min.
Glass bottle
98%
C36H30Cl2NiP2
[(C6H5)3P]2NiCl2
MFCD00009592
02,110; 09,147; 11,165; 16,118
Dichlorobis(triphenylphosphine)nickel(II)
Videos
SDS