missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl) malonate, 98%, Thermo Scientific Chemicals

Catalog Number 10299793
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
18457-04-0
C9H20O4Si2
248.43
MFCD00009631
ATCKJLDGNXGLAO-UHFFFAOYSA-N
bis trimethylsilyl malonate, malonic acid bis trimethylsilyl ester, propanedioic acid, bis trimethylsilyl ester, malonic acid, bis trimethylsilyl ester, malonic acid ditms, malonic acid 2tms, malonic acid, bis-tms, bis trimethylsilyl malonat, malonic acid bis-tms, acmc-1c3vy
87656
bis(trimethylsilyl) propanedioate
C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
18457-04-0
C9H20O4Si2
248.43
MFCD00009631
ATCKJLDGNXGLAO-UHFFFAOYSA-N
bis trimethylsilyl malonate, malonic acid bis trimethylsilyl ester, propanedioic acid, bis trimethylsilyl ester, malonic acid, bis trimethylsilyl ester, malonic acid ditms, malonic acid 2tms, malonic acid, bis-tms, bis trimethylsilyl malonat, malonic acid bis-tms, acmc-1c3vy
87656
bis(trimethylsilyl) propanedioate
C[Si](C)(C)OC(=O)CC(=O)O[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Straw
Density 0.9720g/mL
Boiling Point 63°C to 66°C (1.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiOC(O)CH2C(O)OSi(CH3)3
Refractive Index 1.4150 to 1.417
Quantity 10 g
Specific Gravity 0.972
Solubility Information Solubility in water: reacts
Formula Weight 248.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl) malonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'