missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)carbodiimide, 97%, Thermo Scientific Chemicals

Catalog Number p-7081622
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1000-70-0
C7H18N2Si2
186.405
MFCD00051538
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
70473
N,N'-bis(trimethylsilyl)methanediimine
C[Si](C)(C)N=C=N[Si](C)(C)C
This item is not returnable. View return policy
1000-70-0
C7H18N2Si2
186.405
MFCD00051538
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
70473
N,N'-bis(trimethylsilyl)methanediimine
C[Si](C)(C)N=C=N[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -23°C
Density 0.823
Boiling Point 163°C to 164°C
Flash Point 42°C (107°F)
Odor Mild
Linear Formula (CH3)3SiN=C=NSi(CH3)3
Refractive Index 1.434
Quantity 1 g
UN Number UN1993
Beilstein 2042082
Sensitivity Moisture sensitive
Formula Weight 186.41
Percent Purity 97%
Chemical Name or Material Bis (trimethylsilyl)carbodiimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'